Learn More
Hecht Karl™ Glass Rolled-Rim Vial
Product Code. 10339451
Click to view available options
Volume (Metric):
5 mL
10 mL
15 mL
30 mL
Height (Metric):
24 mm
41 mm
48 mm
72 mm
Diameter (Metric) Outer:
24 mm
25 mm
28 mm
No. per Pack:
108
130
154
Unit Size:
Pack of 108
Pack of 130
Pack of 154
Product Code. 10339451
Brand: Hecht Karl™ 42783016
Only null left
Add to basket
Add to basket
Designed with a rolled rim. Karl Hecht™ Glass Vials are ideal for storing small volumes of liquids.
  • Manufactured with glass for optimum visibility
  • Snap cap included
  • Available in various sizes to fit laboratory needs
Specifications
Color Clear
Diameter (Metric) Outer 24 mm
Material Glass
Volume (Metric) 10 mL
No. per Pack 154
Closure Material Plastic
Closure Type Snap Cap
Height (Metric) 41 mm
Quantity 154 Pk.