Learn More
Hecht Karl™ Glass Rolled-Rim Vial
Click to view available options
Volume (Metric):
5 mL
10 mL
15 mL
30 mL
Height (Metric):
24 mm
41 mm
48 mm
72 mm
Diameter (Metric) Outer:
24 mm
25 mm
28 mm
No. per Pack:
108
130
154
Unit Size:
Pack of 108
Pack of 130
Pack of 154
Description
- Manufactured with glass for optimum visibility
- Snap cap included
- Available in various sizes to fit laboratory needs
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 28 mm |
| Material | Glass |
| Volume (Metric) | 30 mL |
| No. per Pack | 108 |
| Closure Material | Plastic |
| Closure Type | Snap Cap |
| Quantity | 108 Pk. |
| Height (Metric) | 72 mm |