Learn More
Resch Glas™ Glass Rolled Rim Vials
Product Code. 10202901
Change view
Click to view available options
Volume (Metric):
3 mL
10 mL
15 mL
20 mL
25 mL
30 mL
40 mL
100 mL
Height (Metric):
31 mm
46 mm
51 mm
53 mm
56 mm
71 mm
76 mm
81 mm
146 mm
Diameter (Metric) Outer:
19 mm
21 mm
22 mm
24 mm
25 mm
27 mm
28 mm
30 mm
34 mm
Unit Size:
Pack of 100
Pack of 200
Pack of 250
Product Code. Volume (Metric) Height (Metric) Diameter (Metric) Outer unitSize
10202901 15 mL 53 mm 24 mm Pack of 200
10598141 3 mL 31 mm 19 mm Pack of 200
10644141 10 mL 46 mm 22 mm Pack of 200
10252801 10 mL 51 mm 21 mm Pack of 200
10252991 20 mL 56 mm 27 mm Pack of 200
10438312 20 mL 71 mm 25 mm Pack of 200
10418312 25 mL 51 mm 30 mm Pack of 250
10364641 30 mL 76 mm 28 mm Pack of 200
10588911 40 mL 81 mm 30 mm Pack of 200
10598911 100 mL 146 mm 34 mm Pack of 100
10 options
Product Code. 10202901
Brand: Resch Glas™ 9400235
Only null left
Add to basket
Add to basket
Glass Rolled Rim Vials are ideal for a variety of laboratory applications. The vials are made of soda-lime glass with a tight closing polyethylene snap-on lid, non-sterile.
  • Soda-lime glass
  • PE snap-on lid
  • Non-sterile
Specifications
Closure Color Natural
Description Rolled rim glass vials with snap-on lid
Diameter (English) Outer 0.9 in.
Color Clear
Diameter (Metric) Outer 24 mm
Material Soda Lime Glass
Sterility Non-sterile
Volume (Metric) 15 mL
For Use With (Application) A variety of laboratory applications
Height (English) 2.1 in.
Material (Seal) Polyethylene
Neck Style Rolled Rim
Volume (English) 0.5 oz.
Closure Material Polyethylene
Closure Type Snap Cap
Quantity 200 Pk.
Height (Metric) 53 mm
Product Type Rolled Rim Vial