Learn More
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser
Product Code. 10242371
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
20 to 100 mL
40 to 200 mL
6 to 30 mL
Unit Size:
Each
Product Code. 10242371
Brand: Poulten Graf™ 101 08151
Only null left
Add to basket
Add to basket
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
For dispensing of pure organic solvents. Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • Solid dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Product Type Fortuna Optifix Solvent Dispenser
Material PTFE, Polypropylene, Fiberglass
Volume (Metric) 20 to 100 mL
For Use With (Application) For dispensing solvents
Quantity Each