Learn More
Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Description
- For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
- Regular (straight, without notch)
- Supplied in packs of 2
Specifications
Specifications
| For Use With (Equipment) | SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit |
| Type | Low Fluorescence Glass Plate |
| Material | Glass |
| Quantity | 2/Pk. |
| Dimensions (L x W) | 18 x 16 cm |
| Length (Metric) | 18 cm |
| Width (Metric) | 16 cm |