Learn More
Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Product Code. 11519753
Change view
Click to view available options
Type:
Glass Plate
Low Fluorescence Glass Plate
Unit Size:
Pack of 2
Product Code. Type unitSize
11519753 Glass Plate Pack of 2
11905368 Low Fluorescence Glass Plate Pack of 2
2 options
Product Code. 11519753
Brand: Hoefer™ SE6102
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
  • For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
  • Regular (straight, without notch)
  • Supplied in packs of 2
Specifications
For Use With (Equipment) SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Type Glass Plate
Material Glass
Quantity 2/Pk.
Dimensions (L x W) 18 x 16 cm
Length (Metric) 18 cm
Width (Metric) 16 cm