Learn More
Cytiva Glass Plate for Multiphor ll Electrophoresis System
Product Code. 10477304 Shop All Cytiva Products
Change view
Click to view available options
Thickness (Metric):
0.1 cm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) unitSize
10477304 0.1 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10477304 Supplier Cytiva Supplier No. 80110669
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Glass plate for electrophoresis
For use with Multiphor ll Electrophoresis System
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Glass Plate
Material Glass
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Length (Metric) 8.4 cm
Width (Metric) 9.4 cm