Learn More
Socorex™ Glass Pasteur Pipettes
Product Code. 15310965
Change view
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15310965 2 mL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15310965 Supplier Socorex™ Supplier No. 313.02.150
Estimated Shipment: 04-12-2026
Reach into deep vials easily using a long Pasteur pipette. Socorex™ Glass Pasteur Pipettes are supplied non-sterile.
  • General purpose glass Pasteur pipette.
  • Non-sterile.
  • Fit macropipettes when used with appropriate adapter nozzle.
Specifications
Aspirator Plug No
Diameter (Metric) Outer 7 mm
Material Glass
Quantity 250/Pk.
Volume (Metric) 2 mL
Length (Metric) 150 mm
Type Pasteur Pipet
For Use With (Equipment) Acura™ Manual 835.02, Acura™ Electro 936.02, Calibra™ Digital 832.02, Acura™ Manual 835.05, Acura™ Electro 936.05 Macropipettes