Learn More
Hirschmann™ Pasteur Pipettes
Product Code. 10044440
Change view
Click to view available options
Length (Metric):
150 mm
230 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10044440 150 mm Pack of 1000
10400201 230 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10044440 Supplier Hirschmann™ Supplier No. 9250101
  • ONLINE EXCLUSIVE PRICE
    884.29 NOK
    799.67 NOK / Pack of 1000
    Save 84.62 NOK
    9% Off
Estimated Shipment: 03-12-2026
Ideal for transferring samples to sample cups and for drop counting. Hirschmann Instruments™ Pasteur Pipettes are available in either long or short form.
  • Acc. to ISO 7712
  • Without cotton plug
  • With extended fine tip
Specifications
Certifications/Compliance ISO 7712
Material Glass
Includes Finely tapered end
Length (Metric) Tip 40 mm
Quantity 1000 Pack
Top Style Fine drawn
Volume (Metric) 2 mL
Length (Metric) 150 mm
Type Pasteur Pipet