Learn More
Macherey-Nagel™ Glass N9/N11 Wide Opening Micro Insert
Product Code. 12490450 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12490450 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12490450 Supplier Macherey-Nagel™ Supplier No. 702818
  • ONLINE EXCLUSIVE PRICE
    687.00 NOK
    649.56 NOK / Pack of 100
    Save 37.44 NOK
    5% Off
View all promotions
In Stock
Specifications
Color Clear
Material Glass
Volume (Metric) 0.1 mL
Includes With plastic spring
Base Style Conical
Quantity 100 Pk.
For Use With (Equipment) Wide Opening Vials
Height (Metric) 29 mm