Learn More
Macherey-Nagel™ Insert for Small Opening Vials, With Plastic Spring
Product Code. 10093623 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10093623 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10093623 Supplier Macherey-Nagel™ Supplier No. 702824
  • ONLINE EXCLUSIVE PRICE
    687.00 NOK
    649.56 NOK / Pack of 100
    Save 37.44 NOK
    5% Off
View all promotions
In Stock
Use with N8 Screw Neck, N11 Crimp Neck or other small neck vials. Macherey-Nagel™ Insert for Small Opening Vials, With Plastic Spring are 0.1mL glass inserts with a plastic spring that acts as a shock absorber for analyzing micro-samples using an autosampler.
  • 0.1mL glass insert for small opening autosampler vials
  • Plastic spring protects against damage during insertion or if a needle presses down on the bottom
  • Conical design helps to achieve complete removal of the sample
  • Packaged as 100 pieces in a polyethylene (PE) bag
  • Compatible small mouth vials sold separately
Specifications
Color Clear
Diameter (Metric) Outer 5 mm
Material Glass
Volume (Metric) 0.1 mL
For Use With (Application) Small sample chromatography
Includes With plastic spring
Volume (Metric) Insert 0.1 mL
Base Style Conical
Quantity 100 Pk.
For Use With (Equipment) Small Opening Vials
Height (Metric) 29 mm