Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Description
- Not heparinized
- End-to-end, with minicaps
- Acc. to ISO 9000, CE acc. IVD 98/79 EC
- Color-coded vial cap with capillary dispenser
- Conformity certified, with batch number
Specifications
Specifications
| Accuracy | 0.5% |
| Certifications/Compliance | ISO 7550, CE, IVD 98/79 EC |
| Material | Glass |
| Volume (Metric) | 3 μL |
| Length (Metric) | 30 to 32 mm |
| Type | Minicap Capillary Pipette |
| Disposable | Single-use |