Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Product Code. 10666751
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
2 μL
3 μL
4 μL
5 μL
10 μL
20 μL
25 μL
30 μL
40 μL
50 μL
100 μL
Unit Size:
Pack of 1000
Product Code. Volume (Metric) unitSize
10666751 30 μL Pack of 1000
11871060 0.5 μL Pack of 1000
11780055 100 μL Pack of 1000
11871050 10 μL Pack of 1000
11821050 1 μL Pack of 1000
11722293 20 μL Pack of 1000
11881060 25 μL Pack of 1000
11831050 2 μL Pack of 1000
11841050 3 μL Pack of 1000
11851050 4 μL Pack of 1000
11757663 50 μL Pack of 1000
11861050 5 μL Pack of 1000
11732293 40 μL Pack of 1000
13 options
Product Code. 10666751
Brand: Hirschmann™ 9000130
Only null left
Add to basket
Add to basket
Facilitates error-free capillary filling in the laboratory
  • Not heparinized
  • End-to-end, with minicaps
  • Acc. to ISO 9000, CE acc. IVD 98/79 EC
  • Color-coded vial cap with capillary dispenser
  • Conformity certified, with batch number
Specifications
Accuracy 0.5%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material Glass
Volume (Metric) 30 μL
Length (Metric) 30 to 32 mm
Type Minicap Capillary Pipette