Learn More
Glass Microscopic Slides with Ground Edges
Product Code. 13192131
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13192131 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13192131 Supplier Brand Not Defined Supplier No. 2951
Estimated Shipment: 30-11-2026
Use Glass Microscopic Slides with Ground Edges in the laboratory for a clear view of your slide material for analysis and diagnosis.
  • Extra white glass
  • Washed and polished
  • 90 degree ground edges
  • Polished coating
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 90°, Ground
Frosted Yes
Thickness (Metric) 1 to 1.2 mm
Color White
Coating Polished
Quantity 50 Pack