Learn More
Trajan Scientific and Medical™ Glass Lined Stainless-Steel GC Tubing
Exhibits resistance to the harshest of conditions and to strong acids and bases
Brand:  Trajan Scientific and Medical™ 082738
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15209003
  • 860.00 NOK / Each
Estimated Shipment: 04-06-2024
Description
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
Specifications
HPLC Columns, Biological Matrices
1 mm
Borosilicate Glass (Lining)
Tubing
60 cm
Stable up to 500°C