Learn More
Labconco™ Glass Lids for FreeZone™ Freeze Drying Systems
Product Code. 12520857 Shop All Labconco Corporation Products
Change view
Click to view available options
Diameter (English):
14 in.
Weight (English):
3.5 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Weight (English) Unit Size
12520857 14 in. 3.5 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12520857 Supplier Labconco™ Supplier No. 7439902
  • ONLINE EXCLUSIVE PRICE
    10795.00 NOK
    8211.89 NOK / Pack of 1
    Save 2583.11 NOK
    23% Off
 Restricted Item
Estimated Shipment: 18-02-2027
Glass Lids replace the acrylic lid included with FreeZone Freeze Dry Systems.
Glass lid replaces the acrylic lid included with FreeZone 6, 12 and 18 Liter Freeze Dry Systems and with 16-Port Drying Chambers. Provides additional protection from solvents and corrosives that attack acrylic, such as acetonitrile. Includes a neoprene gasket.
Specifications
Type Chamber Lid
Capacity (Metric) 6, 12, 18 L
Material Glass
Size Large
For Use With (Application) Provides additional protection from solvents and corrosives that attach acrylic, such as acetonitrile
For Use With (Equipment) Freezone™ 6, 12 and 18l freeze dry system collector chambers or 16-port drying chamber
Includes 0.25 in. plastic cover and black neoprene gasket
Diameter (English) 14 in.
Diameter (Metric) 35.56 cm
Weight (English) 3.5 lb.
Weight (Metric) 1.6 kg