Learn More
Karl Hecht™ Assistent™ Wide Neck Screw Cap Bottles
Product Code. 11371525
Click to view available options
Capacity (Metric):
25 mL
50 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Color:
Amber
Clear
Unit Size:
Each
Product Code. 11371525
Brand: Karl Hecht™ 41325006
Currently Unavailable: We have no information when this product will be available.
View alternative products
Ideal for the storage of dry powders in the laboratory. Karl Hecht™ Assistent™ Wide Neck Screw Cap Bottles allow you to access their contents with ease.
  • With thread and screw cap
  • Available in clear or amber glass
  • Amber glass is ideal for protecting light-sensitive materials
  • Screw caps are heat resistant up to 110°C
Specifications
Capacity (Metric) 25 mL
Color Amber
Includes Thread and Screw Cap
Material Glass
Mouth Wide
Product Type Jar