Learn More
Hirschmann™ Class AS Graduated Pipettes, Blue Graduation
Product Code. 15231209
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
50 mL
Unit Size:
Pack of 12
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15231209 2 mL Pack of 12
11720834 25 mL Pack of 6
11730834 50 mL Pack of 6
11730814 1 mL Pack of 12
11780824 5 mL Pack of 12
11700834 10 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15231209 Supplier Hirschmann™ Supplier No. 1101141
  • ONLINE EXCLUSIVE PRICE
    1095.00 NOK
    961.64 NOK / Pack of 12
    Save 133.36 NOK
    12% Off
Estimated Shipment: 18-01-2027
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes, Blue Graduation come with a cotton plug end as standard.
  • Acc. to DIN EN ISO 835
  • Main point ring graduation in blue
  • Graduated to tip, zero on top, type 3
  • Conformity certified, with dated batch identification
Specifications
Calibrated Yes
Certifications/Compliance DIN EN ISO 835
Class AS
Material Natural Rubber
For Use With (Application) Pipette filler works via a series of Glass ball valves for air release, suction and delivery
Color Blue
Volume (Metric) 2 mL
Type Pipette Filler
For Use With (Equipment) Pipettes up to 1000 mL