Learn More
Merck Glass Funnel for Vacuum Filtration
Product Code. p-7159153 Shop All Merck Products
Click to view available options
Capacity (Metric):
300 mL
1 L
Description:
Ground glass seal
PTFE-faced
Unit Size:
Each
Product Code. 15869512
Brand: Merck XX1014704
Only null left
Add to basket
Add to basket
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Millipore Glass Vacuum Filtration Accessory ProductThese accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration. This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits, and Millipore MilliSolve Kit.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 47 mm
Capacity (Metric) 300 mL
Material Borosilicate Glass
Autoclavable Not Autoclavable
Color Clear
Description Ground glass seal