Learn More
Glass Filled Nylon Samco Spring Forceps
Product Code. 12760036
Click to view available options
Length (Metric):
125 mm
Unit Size:
Pack of 10
Product Code. 12760036
Brand: S Murray SAMCO L760/01
Only null left
Add to basket
Add to basket
Use these forceps for a variety of applications. S Murray™ Glass Filled Nylon Samco Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
  • 10 per pack
  • Glass filled nylon
  • Autoclavable to 125°C
Specifications
Product Type SAMCO Spring Forceps
Material Glass-filled Nylon
For Use With (Application) For dissection
Quantity 10/Pk.
Tip Style Blunt, Straight
Length (Metric) 125 mm