Learn More
Macherey Nagel Glass Fiber Filter Circles, MN 85/70 BF
Round, glass fiber filters with fast filtration and very high particle retention.
Brand:  Macherey Nagel 404007
Product Code. 10133771
  • ONLINE EXCLUSIVE PRICE
    632.00 NOK
    625.15 NOK / Pack of 100
    Save 6.85 NOK
    1% Off
Only null left
Estimated Shipment: 28-04-2026
Description
  • Manufactured from borosilicate glass fibers
  • Chemically resistant towards most organic and inorganic solvents (except HF)
Specifications
Specifications
70 g/m2
0.6μm
MN 85/70 BF
0.35 mm
70 mm
Round
15 sec.
100/Pk.
70 mm