Learn More
Macherey Nagel Glass Fiber Adhesive Tape
Product Code. 10256331 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 x 50 m reel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10256331 1 x 50 m reel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10256331 Supplier Macherey Nagel Supplier No. 91620
  • ONLINE EXCLUSIVE PRICE
    291.00 NOK
    272.47 NOK / Pack of 1
    Save 18.53 NOK
    6% Off
In Stock
Reinforced tape for securely closing the shipping boxes of hazardous goods.
  • Glass fiber reinforced
Specifications
Product Type Adhesive Tape
Quantity 1 x 50 m reel