Learn More
Jaytec Glass™ Glass Dropping Pipets with Teat
Click to view available options
Unit Size:
Pack of 12
Graduations:
1 mL
Description
Natural rubber cap assists in retrieving and dispensing liquid/media.
Specifications
Specifications
| Material | Glass, Rubber |
| Graduated | No |
| Includes | Teat |
| Quantity | 12 Pack |
| Volume (Metric) | 2 mL |
| Type | Dropping Pipet |