Learn More
Nordic Pack Glass Dropping Pipettes
Product Code. 11341515
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Pack of 114
Pack of 150
Pack of 68
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11341515 100 mL Pack of 68
11321515 30 mL Pack of 150
11311515 10 mL Each
11331515 50 mL Pack of 114
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11341515 Supplier Nordic Pack Supplier No. 11100
Estimated Shipment: 23-11-2026
Glass dropping Pipettes
  • Made from glass
  • Available in different capacities
Specifications
Diameter (Metric) Outer 47 mm
Material Glass
Volume (Metric) 100 mL
Type Dropping Pipette