Learn More
Nordic Pack Glass Dropping Pipettes
Product Code. 11321515
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Pack of 114
Pack of 150
Pack of 68
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11321515 30 mL Pack of 150
11311515 10 mL Each
11341515 100 mL Pack of 68
11331515 50 mL Pack of 114
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11321515 Supplier Nordic Pack Supplier No. 1130
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Glass dropping Pipettes
  • Made from glass
  • Available in different capacities
Specifications
Diameter (Metric) Outer 32 mm
Material Glass
Volume (Metric) 30 mL
Type Dropping Pipette