Learn More
Hirschmann™ Glass covers
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W):
21 x 26 mm
24 x 40 mm
24 x 60 mm
missing translation for 'unitSize'
Pack of 1000
Description
- LOT number on packaging
- CE pursuant to IVD 98/79
- DIN 58 884
Specifications
Specifications
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Dimensions (L x W) | 24 x 60 mm |