Description
- Manufactured from Borosilicate glass
- Complies with ISO 4800 standards
- Supplied with a glass and plastic stopper
- Available with interchangeable glass or PTFE stopcock
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | ISO 4800 |
| Quantity | Each |
| Stopcock | Glass |
| Stopper Material | Glass, Plastic |
| Stopper Number | 29/32 |
| Product Type | Separating funnel |