Specifications
| Base Material | Silica |
| Carbon Load | 8% |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| For Use With (Application) | Pesticides, PCBs Applications |
| Particle Size | 45 μm |
| Type | Solid Phase Extraction Column |
| Endcapped | No |
| Pore Size | 60 μm |
| Surface Area (Metric) | 500 m2/g |