Learn More
DWK Life Sciences Glass Ceramic Laboratory Protection Plate
Product Code. 11708041 Shop All DWK Life Sciences Products
Change view
Click to view available options
Dimensions (L x W):
135 x 135 mm
155 x 155 mm
175 x 175 mm
Unit Size:
Pack of 10
Product Code. Dimensions (L x W) unitSize
11708041 155 x 155 mm Pack of 10
11767991 135 x 135 mm Pack of 10
12540997 175 x 175 mm Pack of 10
3 options
Product Code. 11708041
Brand: DWK Life Sciences 238215703
Only null left
Add to basket
Add to basket
The glass ceramic plates are suitable for heating of cooking glasses with the Bunsen burner.
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • Due to low thermal expansion stresses, well suited to heating glassware with a Bunsen burner
  • Protects cooking glasses from direct flame
TRUSTED_SUSTAINABILITY
Specifications
Material Glass Ceramics
Autoclavable Not Autoclavable
Dimensions (L x W) 155 x 155 mm
For Use With (Application) Heating glassware; Square Quadrupod for Glass Ceramic Laboratory Protection Plate
No. per Pack 10/Pk.