Learn More
Karl Hecht™ Glass Centrifuge Tubes, Screw Cap
Click to view available options
Capacity (Metric):
10 to 15 mL
Unit Size:
Pack of 100
Description
- Threaded tube with cap closures.
- Ungraduated.
- Sterilizable up to 121°C.
- Conical bottoms.
- AR™ glass construction.
Specifications
Specifications
| Capacity (Metric) | 10 to 15 mL |
| Material | Soda Lime Glass |
| For Use With | Centrifuge |
| No. per Pack | 100 |
| Autoclavable | Autoclavable |
| Length (Metric) | 112 mm |
| Bottom Shape | Conical |
| Closure Type | Screw Cap |
| Closure Included | Yes |
| Diameter (Metric) | 17 mm |