Learn More
Karl Hecht™ Glass Centrifuge Tubes, Conical
Description
- Conical bottom.
- Graduated or ungraduated.
- Rimmed or rimless.
- AR™ soda glass, or Duran™ borosilicate.
Specifications
Specifications
| Capacity (Metric) | 10 to 15 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With | Centrifuge |
| No. per Case | 100/Box |
| Length (Metric) | 112 mm |
| Bottom Shape | Conical |
| Diameter (Metric) | 17 mm |
| Graduated | No |
| Product Type | Rimmed Centrifuge Tube |