Learn More
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point
Product Code. p-8009727
Change view
Click to view available options
Length (Metric) Tube:
70 mm
75 mm
80 mm
Diameter (Metric) Inner:
0.42 mm
0.9 mm
1 mm
1.2 mm
1.5 mm
Product Type:
Capillary Tubes (Both Ends Open)
Capillaty Tubes (One End Closed)
Unit Size:
Pack of 1000
Product Code. Length (Metric) Tube Diameter (Metric) Inner Product Type unitSize
10640783 80 mm 1.2 mm Capillary Tubes (Both Ends Open) Pack of 1000
10764034 80 mm 1.2 mm Capillaty Tubes (One End Closed) Pack of 1000
10185413 80 mm 1 mm Capillaty Tubes (One End Closed) Pack of 1000
10222413 80 mm 0.42 mm Capillary Tubes (Both Ends Open) Pack of 1000
10501363 70 mm 0.9 mm Capillary Tubes (Both Ends Open) Pack of 1000
16341238 75 mm 1.5 mm Capillaty Tubes (One End Closed) Pack of 1000
6 options
Product Code. 10640783
Brand: Marienfeld Superior™ 2930207
Use Marienfeld Superior™ Glass Capillary Tubes for storage of small amounts of liquids and to determine melting points during laboratory experimentation.
  • With rectangular cut edges
  • For single use
  • 100 capillaries in a dispenser vial, 10 vials in a box
Specifications
Material Glass
Product Type Capillary Tubes (Both Ends Open)
Diameter (Metric) Inner 1.2 mm
For Use With (Application) For Determination Of Melting Point
Length (Metric) Tube 80 mm
Quantity 1000