Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Description
- With ringmark
- Conformity certified
- Manufactured from durable DURAN™ glass
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
| Accuracy | 0.25% |
| Certifications/Compliance | ISO 7550, CE, IVD 98/79 EC |
| Material | Glass |
| Includes | Ring Mark |
| Volume (Metric) | 20 μL |
| Type | Capillary Pipette |
| Disposable | Single-use |