Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Description
- With polyethylene stopper
- With amber stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Polyethylene |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 300 mm |
| Limit of Error | ±0.4 mL |
| Standard Taper Joint | 24/29 NS Standard Ground |
| Tolerance | ±0.4 mL |
| Graduated | Yes |
| Class | A |