Learn More
Nordic Pack Glass Bottles for Storing Medicines
Product Code. 11351495
Encompass
Change view
Click to view available options
Capacity (Metric):
60 mL
100 mL
300 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 36
Pack of 85
Product Code. Capacity (Metric) unitSize
11351495 100 mL Each
11341495 60 mL Pack of 85
11371495 300 mL Pack of 36
11381495 500 mL Each
11391495 1000 mL Each
5 options
Product Code. 11351495
Brand: Nordic Pack 100LV
Only null left
Add to basket
Add to basket
For storing medicines
  • Made from glass
  • Available in different capacities
Specifications
Capacity (Metric) 100 mL
Color Brown
Diameter (Metric) Outer Neck 47 mm
Material Glass
Thread Size (Metric) 28 mm
Closure Included No
Mouth Narrow
Height (Metric) 112 mm