Learn More
Gravis Wide Neck Glass Bottles
Product Code. p-8000722
Change view
Click to view available options
Capacity (Metric):
2.5 mL
30 mL
100 mL
Unit Size:
Pack of 110
Pack of 360
Pack of 86
Product Code. Capacity (Metric) unitSize
11864282 2.5 mL Pack of 360
12646326 30 mL Pack of 110
17624815 100 mL Pack of 86
3 options
Product Code. 11864282
Brand: Gravis A001021110
Provides excellent storage for liquids and solids in the laboratory. Gravis Wide Neck Glass Bottles are to be sealed with waterproof screw caps.
  • Available in a range of capacities
  • Wide neck allows for ease of access to contents
  • Manufactured from soda lime glass
Specifications
Capacity (Metric) 2.5 mL
Material Glass
No. per Pack 360/pk
Quantity 2.5mL
Product Type Storage Bottle