Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Product Code. 11782412
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
7/16 NS Standard Taper Ground
Tolerance:
±0.025 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
±1.2 mL
±2 mL
0.025 mL
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Standard Taper Joint Tolerance unitSize
11782412 5 mL 7/16 NS Standard Taper Ground 0.025 mL Pack of 2
10218211 10 L 45/40 NS Standard Ground ±2 mL Each
10689451 1000 mL 24/29 NS Standard Ground ±0.6 mL Each
10522891 1000 mL 24/29 NS Standard Ground ±0.4 mL Each
11780964 100 mL 12/21 NS Standard Ground ±0.1 mL Pack of 2
11760964 10 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11708071 10 mL 7/16 NS Standard Ground ±0.025 mL Pack of 2
10561161 2000 mL 29/32 NS Standard Ground ±0.6 mL Each
11782142 200 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11718071 20 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11792142 250 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11816993 25 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
11728071 25 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
10244712 20 mL 12/21 NS Standard Ground ±0.04 mL Pack of 2
11702152 500 mL 19/26 NS Standard Ground ±0.25 mL Pack of 2
10646111 5 L 34/35 NS Standard Ground ±1.2 mL Each
11770964 50 mL 14/23 NS Standard Ground ±0.08 mL Pack of 2
11750964 5 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11738071 50 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
11772142 100 mL 14/23 NS Standard Ground ±0.1 mL Pack of 2
20 options
Product Code. 11782412
Brand: Hirschmann™ 2820152
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
  • With NS ground and polyethylene stopper
  • Acc. to DIN ENO ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 5 mL
Material Borosilicate Glass
Height (Metric) 70 ± 5 mm
Standard Taper Joint 7/16 NS Standard Taper Ground
Tolerance 0.025 mL
Graduated Yes
Class A