Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Product Code. 11750964
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
7/16 NS Standard Taper Ground
Tolerance:
±0.025 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
±1.2 mL
±2 mL
0.025 mL
Unit Size:
Each
Pack of 2
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Tolerance unitSize
11750964 5 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
10218211 10 L 45/40 NS Standard Ground ±2 mL Each
10689451 1000 mL 24/29 NS Standard Ground ±0.6 mL Each
10522891 1000 mL 24/29 NS Standard Ground ±0.4 mL Each
11780964 100 mL 12/21 NS Standard Ground ±0.1 mL Pack of 2
11760964 10 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11708071 10 mL 7/16 NS Standard Ground ±0.025 mL Pack of 2
10561161 2000 mL 29/32 NS Standard Ground ±0.6 mL Each
11782142 200 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11718071 20 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11792142 250 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11816993 25 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
11728071 25 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
10244712 20 mL 12/21 NS Standard Ground ±0.04 mL Pack of 2
11702152 500 mL 19/26 NS Standard Ground ±0.25 mL Pack of 2
10646111 5 L 34/35 NS Standard Ground ±1.2 mL Each
11770964 50 mL 14/23 NS Standard Ground ±0.08 mL Pack of 2
11738071 50 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
11772142 100 mL 14/23 NS Standard Ground ±0.1 mL Pack of 2
11782412 5 mL 7/16 NS Standard Taper Ground 0.025 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 11750964 Supplier Hirschmann™ Supplier No. 2820153
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 5 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 70 mm
Limit of Error ±0.04 mL
Standard Taper Joint 10/19 NS Standard Ground
Tolerance ±0.04 mL
Graduated Yes
Class A