Learn More
Velp Scientifica™ Glass Beakers
Product Code. 15359624 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15359624 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15359624 Supplier Velp Scientifica™ Supplier No. A00001001
Excellent for holding solutions during the flocculation process. Velp Scientifica™ Glass Beakers are for use with FC S and JLT Series Flocculators.
  • Excellent chemical resistance
  • Easy to use
  • Clear glass
  • Graduation marks
Specifications
Capacity (Metric) 1000 mL
For Use With (Equipment) FC S and JLT Series Flocculators
Type Beaker