Learn More
RETSCH Glass Beads for Mills
Product Code. 10262641 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
0.10 to 0.25 mm
0.25 to 0.50 mm
0.50 to 0.75 mm
Unit Size:
500g
Product Code. Diameter (Metric) unitSize
10262641 0.50 to 0.75 mm 500g
10069270 0.10 to 0.25 mm 500g
10252641 0.25 to 0.50 mm 500g
3 options
Product Code. 10262641
Brand: RETSCH 22.222.0003
Only null left
Add to basket
Add to basket
For use with Retsch MM 200 Mixer Mills.
  • Ideal for disruption of biological cells
Specifications
Type Beads
Material Glass
Diameter (Metric) 0.50 to 0.75 mm
For Use With (Equipment) Mixer Mill MM 200
Quantity 500 g