Learn More
Hecht Karl™ Glass Assistant™ Fermenting Tubes
Product Code. 10318721
Change view
Click to view available options
Diameter (Metric) Outer:
10 to 11 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10318721 10 to 11 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10318721 Supplier Hecht Karl™ Supplier No. 41350010
  • ONLINE EXCLUSIVE PRICE
    69.23 NOK
    36.83 NOK / Pack of 1
    Save 32.40 NOK
    46% Off
Estimated Shipment: 12-01-2027
Specifications
Material Glass
Diameter (Metric) Outer 10 to 11 mm
Overall Length (Metric) 200 mm
Packaging Quantity 1 Pack
Product Type Fermenting Tube