Learn More
Thermo Scientific™ GLACIER Series G50 Ultra-Low Refrigerated Circulators
These Ultra-Low Refrigerated Circulators are cost effective with extreme temperature performance, offering high heating and cooling capacities for rapid heat-up and cool-down.
Description
Description
The Thermo Scientific™ GLACIER Series G50 Ultra-Low Refrigerated Circulators are cost effective and offer extreme temperature performance. These circulators deliver high heating and cooling capacities for rapid heat-up and cool-down times. Fitted with adjustable castors, drainport and handles, it's a perfect fit for any environment. Available heated top plate prevents condensation and icing at low temperatures.
  • Suitable for external temperature applications with simultaneous operation in the bath tank
  • Deliver high heating and cooling capacities for rapid heatup and cool down times
  • Fitted with adjustable casters, drain port, and handle
  • Heated top plate prevents condensation and icing at low temperatures
Includes:
Communication cables, bridge with gasket & thumbscrews, 8 and 12mm hose barbs with clamps, 6ft. power cord, sealed work area cover, 3-year warranty
Recommended for:
  • Jacketed reaction vessels
  • Material testing
  • Analytical instrumentation
  • Calibration
  • Condensers
  • Crystallization
  • Warranty varies by country, please contact us for details