missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (109114) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6942JF669
4687.75 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Specifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18, Glu52-Ser177, Accession # Q9UNG2 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 109114 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis | |
| 8995 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction