missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIT1 Antibody (924640) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8508JF669
This item is not returnable.
View return policy
Description
GIT1 Monoclonal antibody specifically detects GIT1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| GIT1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 924640 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 28964 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction