missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18034-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.Specifications
GIP | |
Polyclonal | |
Unconjugated | |
GIP | |
gastric inhibitory polypeptide, GIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2695 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09681 | |
GIP | |
GIP Fusion Protein Ag12634 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |