missing translation for 'onlineSavingsMsg'
Learn More

GIP Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18034-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898064

  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.
Specifications

Specifications

GIP
Polyclonal
Unconjugated
GIP
gastric inhibitory polypeptide, GIP
Rabbit
Antigen Affinity Chromatography
RUO
2695
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09681
GIP
GIP Fusion Protein Ag12634
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers