missing translation for 'onlineSavingsMsg'
Learn More

Ginsenoside Rh1, MedChemExpress

Product Code. p-200095998
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16439038

Brand: Medchem Express HYN0604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ginsenoside Rh1 (Prosapogenin A2) inhibits the expression of PPAR-γ, TNF-α, IL-6, and IL-1β.

Ginsenoside Rh1 (Prosapogenin A2) inhibits the expression of PPAR-γ, TNF-α, IL-6, and IL-1β.

Specifications

Chemical Name or Material Ginsenoside Rh1
CAS 63223-86-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H62O9
Quantity 10 mg
Synonym Prosapogenin A2 Sanchinoside B2 Sanchinoside Rh1
Solubility Information DMSO : 100 mg/mL (156.53 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(C[C@H](O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)[C@@]4([H])C5(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC2)([H])[C@]4(CC[C@@H]5O)C
Molecular Weight (g/mol) 638.87
Formula Weight 638.87
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.