Learn More
Ginsenoside Rh1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ginsenoside Rh1 (Prosapogenin A2) inhibits the expression of PPAR-γ, TNF-α, IL-6, and IL-1β.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rh1 |
| CAS | 63223-86-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H62O9 |
| Quantity | 10 mg |
| Synonym | Prosapogenin A2 Sanchinoside B2 Sanchinoside Rh1 |
| Solubility Information | DMSO : 100 mg/mL (156.53 mM; Need ultrasonic) |
| SMILES | C[C@@]([C@@]12C)(C[C@H](O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)[C@@]4([H])C5(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC2)([H])[C@]4(CC[C@@H]5O)C |
| Molecular Weight (g/mol) | 638.87 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.