Learn More
GINS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15651-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein subunit of the GINS heterotetrameric complex, which is essential for the initiation of DNA replication and replisome progression in eukaryotes. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| GINS3 | |
| Polyclonal | |
| Unconjugated | |
| GINS3 | |
| GINS complex subunit 3, GINS3, PSF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64785, 78833 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BRX5, Q9CY94 | |
| GINS3 | |
| GINS3 Fusion Protein Ag8247 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |