missing translation for 'onlineSavingsMsg'
Learn More

GILZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12352-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850024

  • 1415.13 NOK / 20µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.
Specifications

Specifications

GILZ
Polyclonal
Unconjugated
TSC22D3
DIP, DSIP immunoreactive peptide, DSIPI, GILZ, hDIP, Protein DIP, TSC 22 like protein, TSC 22 related protein, TSC 22R, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC22D3
Rabbit
Antigen Affinity Chromatography
RUO
14605, 1831, 83514
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99576, Q9EQZ1, Q9Z2S7
TSC22D3
GILZ Fusion Protein Ag3015
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers